Assay Details
Target Gene Details
Entrez Gene ID: | 91408 |
Gene Name: | basic transcription factor 3 like 4 |
Gene Aliases: |
- |
Location: |
Chr.1:52056185-52090716 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BTF3L4 | NM_001136497.2 | 5 | 2325 | NP_001129969.1 |
| NM_001243767.1 | 5 | 2190 | NP_001230696.1 | |
| NM_152265.4 | 6 | 2392 | NP_689478.1 | |
| AK027582.1 | ||||
| BC021004.1 | AAH21004.1 |
More Information
Set Membership: |
|

Genomic Map