Assay Details
Target Gene Details
Entrez Gene ID: | 284541 |
Gene Name: | cytochrome P450 family 4 subfamily A member 22 |
Gene Aliases: |
- |
Location: |
Chr.1:47137425-47149738 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP4A22 | NM_001010969.3 | NP_001010969.2 | ||
| NM_001308102.1 | NP_001295031.1 | |||
| XM_005270767.3 | XP_005270824.1 | |||
| XM_005270770.3 | XP_005270827.1 | |||
| AY280371.1 | AAQ21367.1 | |||
| AY280372.1 | AAQ21368.1 | |||
| BC142607.1 | ||||
| BC142690.1 | ||||
| BC148248.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv945929 | Chr.1:47113079 - 47151823 on Build GRCh38 | Duplication |
|
| esv2763300 | Chr.1:46892860 - 47163102 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map