Assay Details
Target Gene Details
Entrez Gene ID: | 114803 |
Gene Name: | Myb like, SWIRM and MPN domains 1 |
Gene Aliases: |
2A-DUB, 2ADUB |
Location: |
Chr.1:58654739-58700091 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYSM1 | NM_001085487.2 | 20 | 3702 | NP_001078956.1 |
| XM_006710314.3 | 20 | 3715 | XP_006710377.1 | |
| XM_011540573.2 | 21 | 3682 | XP_011538875.1 | |
| XM_011540574.2 | 20 | 3950 | XP_011538876.1 | |
| XM_017000195.1 | 21 | 3754 | XP_016855684.1 | |
| XM_017000196.1 | 18 | 3604 | XP_016855685.1 | |
| XM_017000197.1 | 16 | 3382 | XP_016855686.1 | |
| AB067502.1 | 16 | 3728 | BAB67808.1 | |
| BX537912.1 | 7 | 1966 | CAD97896.1 | |
| CR749450.1 | 3 | 2563 | CAH18287.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003930 | Chr.1:58464772 - 58691366 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78520326] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map