Assay Details
Target Gene Details
Entrez Gene ID: | 81569 |
Gene Name: | actin like 8 |
Gene Aliases: |
CT57 |
Location: |
Chr.1:17755313-17827063 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTL8 | NM_030812.2 | NP_110439.2 | ||
| XM_011542212.2 | XP_011540514.1 | |||
| AK057339.1 | BAB71434.1 | |||
| BC028909.1 | AAH28909.1 | |||
| BI826094.1 | ||||
| U20582.1 | AAA62476.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508248 | Chr.1:17737910 - 17813810 on Build GRCh38 | Deletion |
|
| nsv1160182 | Chr.1:17655418 - 18282385 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72927118] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map