Assay Details
Target Gene Details
Entrez Gene ID: | 148423 |
Gene Name: | chromosome 1 open reading frame 52 |
Gene Aliases: |
gm117 |
Location: |
Chr.1:85249953-85259672 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1orf52 | NM_198077.3 | NP_932343.1 | ||
| NR_024113.1 | ||||
| AK126929.1 | ||||
| AY486331.1 | AAS57906.1 | |||
| AY486332.1 | AAS57907.1 | |||
| BC029538.1 | AAH29538.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34165 | Chr.1:85076357 - 85380566 on Build GRCh38 | Loss |
|
| nsv830459 | Chr.1:85123902 - 85320628 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map