Assay Details
Target Gene Details
Entrez Gene ID: | 6282 |
Gene Name: | S100 calcium binding protein A11 |
Gene Aliases: |
HEL-S-43, MLN70, S100C |
Location: |
Chr.1:152032506-152037035 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100A11 | NM_005620.1 | NP_005611.1 | ||
| BC001410.2 | AAH01410.1 | |||
| BC014354.1 | AAH14354.1 | |||
| D38583.1 | BAA07597.1 | |||
| D49355.1 | BAA08354.1 | |||
| D50374.1 | BAA23325.1 | |||
| EU794659.1 | ||||
| X80201.1 | CAA56492.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1007097 | Chr.1:151860320 - 152255501 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map