Assay Details
Target Gene Details
Entrez Gene ID: | 84288 |
Gene Name: | EF-hand calcium binding domain 2 |
Gene Aliases: |
CFAP200, DRC8 |
Location: |
Chr.1:244969739-245125228 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFCAB2 | NM_001143943.1 | NP_001137415.1 | ||
| NM_001290327.1 | NP_001277256.1 | |||
| NM_032328.3 | NP_115704.1 | |||
| NR_026587.1 | ||||
| XM_011544300.2 | XP_011542602.1 | |||
| XM_011544301.1 | XP_011542603.1 | |||
| XM_011544302.2 | XP_011542604.1 | |||
| XM_017002545.1 | 1 | 348 | XP_016858034.1 | |
| AK001393.1 | ||||
| AK090927.1 | ||||
| AK302883.1 | ||||
| BC005357.1 | AAH05357.1 | |||
| BG546306.1 | ||||
| BG701985.1 | ||||
| BI838837.1 | ||||
| BQ438351.1 | ||||
| DA156641.1 | ||||
| DB286332.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3589420 | Chr.1:244890996 - 245040215 on Build GRCh38 | Loss |
|
| nsv5188 | Chr.1:244962548 - 244994285 on Build GRCh38 | Insertion |
|
| esv22057 | Chr.1:244894679 - 245033598 on Build GRCh38 | Loss |
|
| dgv602n100 | Chr.1:244936197 - 245539982 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map