Assay Details
Target Gene Details
Entrez Gene ID: | 8564 |
Gene Name: | kynurenine 3-monooxygenase (kynurenine 3-hydroxylase) |
Gene Aliases: |
dJ317G22.1 |
Location: |
Chr.1:241531883-241595647 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KMO | NM_003679.4 | NP_003670.2 | ||
| XM_005273337.1 | XP_005273394.1 | |||
| XM_005273338.3 | XP_005273395.1 | |||
| XM_017002649.1 | XP_016858138.1 | |||
| AF056032.1 | AAC62615.1 | |||
| AK291558.1 | ||||
| AK310937.1 | ||||
| AU137977.1 | ||||
| BC005297.1 | AAH05297.1 | |||
| BP276365.1 | ||||
| Y13153.1 | CAA73613.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525983 | Chr.1:241543233 - 241548401 on Build GRCh38 | Loss |
|
| nsv549460 | Chr.1:241331278 - 241669985 on Build GRCh38 | Gain |
|
| nsv1002214 | Chr.1:241295639 - 241936488 on Build GRCh38 | Gain |
|
| dgv159e212 | Chr.1:241544439 - 241556478 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map