Assay Details
Target Gene Details
Entrez Gene ID: | 93185 |
Gene Name: | immunoglobulin superfamily member 8 |
Gene Aliases: |
CD316, CD81P3, EWI-2, EWI2, KCT-4, LIR-D1, PGRL |
Location: |
Chr.1:160091339-160099442 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGSF8 | NM_001206665.2 | NP_001193594.1 | ||
| NM_001320247.1 | NP_001307176.1 | |||
| NM_052868.5 | NP_443100.1 | |||
| XM_006711631.3 | XP_006711694.1 | |||
| XM_011510159.2 | 1 | 586 | XP_011508461.1 | |
| XM_017002835.1 | XP_016858324.1 | |||
| XM_017002836.1 | XP_016858325.1 | |||
| XM_017002837.1 | XP_016858326.1 | |||
| XM_017002840.1 | XP_016858329.1 | |||
| AF311906.1 | AAM94901.1 | |||
| AF407274.1 | AAL01052.1 | |||
| AK055843.1 | BAB71027.1 | |||
| AK308115.1 | ||||
| AY044845.1 | AAK92220.1 | |||
| AY157579.1 | AAO13163.1 | |||
| BC053881.1 | AAH53881.1 | |||
| BM544492.1 | ||||
| DB501388.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952222 | Chr.1:160082711 - 160099010 on Build GRCh38 | Deletion |
|
| nsv520189 | Chr.1:160041722 - 160125835 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map