Assay Details
Target Gene Details
Entrez Gene ID: | 1999 |
Gene Name: | E74 like ETS transcription factor 3 |
Gene Aliases: |
EPR-1, ERT, ESE-1, ESX |
Location: |
Chr.1:202010519-202017188 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELF3 | NM_001114309.1 | 9 | 3133 | NP_001107781.1 |
| NM_004433.4 | 9 | 3019 | NP_004424.3 | |
| BQ023145.1 | 1 | 131 | ||
| BX093212.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516812 | Chr.1:202008699 - 202017290 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map