Assay Details
Target Gene Details
Entrez Gene ID: | 3752 |
Gene Name: | potassium voltage-gated channel subfamily D member 3 |
Gene Aliases: |
BRGDA9, KCND3L, KCND3S, KSHIVB, KV4.3, SCA19, SCA22 |
Location: |
Chr.1:111770662-111989577 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCND3 | XM_005270851.4 | 8 | 5116 | XP_005270908.1 |
| XM_006710629.3 | 8 | 5426 | XP_006710692.1 | |
| XM_006710630.3 | 7 | 5059 | XP_006710693.1 | |
| XM_017001244.1 | 8 | 5441 | XP_016856733.1 |
More Information
Set Membership: |
|

Genomic Map