Assay Details
Target Gene Details
Entrez Gene ID: | 2140 |
Gene Name: | EYA transcriptional coactivator and phosphatase 3 |
Gene Aliases: |
- |
Location: |
Chr.1:27970344-28088671 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EYA3 | NM_001282560.1 | NP_001269489.1 | ||
| NM_001282561.1 | NP_001269490.1 | |||
| NM_001282562.1 | NP_001269491.1 | |||
| NM_001990.3 | NP_001981.2 | |||
| XM_006710449.3 | XP_006710512.1 | |||
| XM_011540999.2 | XP_011539301.1 | |||
| XM_011541001.2 | XP_011539303.1 | |||
| XM_011541003.2 | XP_011539305.1 | |||
| XM_011541004.2 | XP_011539306.1 | |||
| AK289805.1 | ||||
| AK295745.1 | ||||
| AK298129.1 | ||||
| BC041667.1 | AAH41667.1 | |||
| BX648945.1 | ||||
| U81602.1 | AAB42066.1 |
More Information
Set Membership: |
|

Genomic Map