Assay Details
Target Gene Details
Entrez Gene ID: | 6421 |
Gene Name: | splicing factor proline and glutamine rich |
Gene Aliases: |
POMP100, PPP1R140, PSF |
Location: |
Chr.1:35176378-35193161 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SFPQ | NR_136702.1 | |||
| NR_136703.1 | ||||
| XM_005271112.4 | 10 | 4199 | XP_005271169.1 | |
| XM_005271113.4 | 10 | 4199 | XP_005271170.1 | |
| XM_005271115.4 | XP_005271172.1 | |||
| XM_011541950.2 | 10 | 4253 | XP_011540252.1 | |
| XM_017002053.1 | 10 | 4199 | XP_016857542.1 | |
| XM_017002054.1 | 10 | 4199 | XP_016857543.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs114776206,rs142590388] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map