Assay Details
Target Gene Details
Entrez Gene ID: | 164091 |
Gene Name: | progestin and adipoQ receptor family member 7 |
Gene Aliases: |
MPRA, PGLP, mSR |
Location: |
Chr.1:25861484-25876707 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAQR7 | XM_011540861.1 | XP_011539163.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv460884 | Chr.1:25778482 - 25891593 on Build GRCh38 | Loss |
|
| nsv834491 | Chr.1:25726411 - 25914117 on Build GRCh38 | Loss |
|
| nsv834502 | Chr.1:25834281 - 25987428 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map