Assay Details
Target Gene Details
Entrez Gene ID: | 2140 |
Gene Name: | EYA transcriptional coactivator and phosphatase 3 |
Gene Aliases: |
- |
Location: |
Chr.1:27970344-28088671 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
More Information
Set Membership: |
|

Genomic Map