Assay Details
Target Gene Details
Entrez Gene ID: | 11123 |
Gene Name: | RCAN family member 3 |
Gene Aliases: |
DSCR1L2, MCIP3, RCN3, hRCN3 |
Location: |
Chr.1:24502351-24537020 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RCAN3 | NM_001251977.1 | NP_001238906.1 | ||
| NM_001251978.1 | NP_001238907.1 | |||
| NM_001251979.1 | NP_001238908.1 | |||
| NM_001251980.1 | NP_001238909.1 | |||
| NM_001251981.1 | NP_001238910.1 | |||
| NM_001251982.1 | NP_001238911.1 | |||
| NM_001251983.1 | NP_001238912.1 | |||
| NM_001251984.1 | NP_001238913.1 | |||
| NM_001251985.1 | NP_001238914.1 | |||
| NM_013441.3 | NP_038469.1 | |||
| AF176116.1 | AAF01684.1 | |||
| AF176117.1 | AAF01685.1 | |||
| AY906854.1 | AAW83514.1 | |||
| BC035854.1 | AAH35854.1 | |||
| BP326714.1 | ||||
| CT001954.1 | ||||
| CX752875.1 | ||||
| EF431960.1 | ||||
| EF467309.1 | ||||
| EF529733.1 | ||||
| GQ411200.1 | ||||
| HQ287726.1 | ||||
| HQ317421.1 | ||||
| HQ317422.1 | ||||
| HQ317423.1 | ||||
| HQ317426.1 | ||||
| JN203053.1 | ||||
| JN228252.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs114383419] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map