Assay Details
Target Gene Details
Entrez Gene ID: | 6518 |
Gene Name: | solute carrier family 2 member 5 |
Gene Aliases: |
GLUT-5, GLUT5 |
Location: |
Chr.1:9036941-9093407 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3585161 | Chr.1:9045371 - 9049438 on Build GRCh38 | Gain |
|
| nsv1004025 | Chr.1:9000983 - 9069086 on Build GRCh38 | Gain |
|
| esv3577721 | Chr.1:9044393 - 9059929 on Build GRCh38 | Loss |
|
| nsv545347 | Chr.1:9020352 - 9072997 on Build GRCh38 | Loss |
|
| nsv831981 | Chr.1:9031658 - 9187508 on Build GRCh38 | Loss |
|
| dgv15n100 | Chr.1:8992095 - 9056684 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map