Assay Details
Target Gene Details
Entrez Gene ID: | 200010 |
Gene Name: | solute carrier family 5 member 9 |
Gene Aliases: |
SGLT4 |
Location: |
Chr.1:48221113-48248644 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC5A9 | XM_011540924.2 | 1 | 1136 | XP_011539226.1 |
| XM_011540925.2 | 1 | 1137 | XP_011539227.1 | |
| XM_011540929.2 | 1 | 1141 | XP_011539231.1 | |
| XM_017000558.1 | 1 | 1140 | XP_016856047.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3578249 | Chr.1:48202315 - 48332128 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map