Assay Details
Target Gene Details
Entrez Gene ID: | 400823 |
Gene Name: | family with sequence similarity 177 member B |
Gene Aliases: |
- |
Location: |
Chr.1:222737216-222772193 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM177B | NM_001324080.1 | NP_001311009.1 | ||
| NM_207468.2 | NP_997351.2 | |||
| NR_136691.1 | ||||
| XM_017001279.1 | XP_016856768.1 | |||
| XM_017001280.1 | XP_016856769.1 | |||
| XM_017001281.1 | XP_016856770.1 | |||
| XM_017001282.1 | XP_016856771.1 | |||
| XM_017001283.1 | XP_016856772.1 | |||
| XM_017001284.1 | XP_016856773.1 | |||
| XM_017001285.1 | XP_016856774.1 | |||
| AK125494.1 | BAC86182.1 | |||
| CA848039.1 |

Genomic Map