Assay Details
Target Gene Details
Entrez Gene ID: | 5825 |
Gene Name: | ATP binding cassette subfamily D member 3 |
Gene Aliases: |
ABC43, CBAS5, PMP70, PXMP1, ZWS2 |
Location: |
Chr.1:94385106-94518663 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCD3 | XM_017001953.1 | XP_016857442.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs72968322] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map