Assay Details
Target Gene Details
Entrez Gene ID: | 5825 |
Gene Name: | ATP binding cassette subfamily D member 3 |
Gene Aliases: |
ABC43, CBAS5, PMP70, PXMP1, ZWS2 |
Location: |
Chr.1:94385106-94518663 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCD3 | NM_001122674.1 | NP_001116146.1 | ||
| NM_002858.3 | NP_002849.1 | |||
| XM_006710802.2 | XP_006710865.2 | |||
| XM_017001953.1 | XP_016857442.1 | |||
| XM_017001954.1 | XP_016857443.1 | |||
| AK296791.1 | ||||
| AK302714.1 | ||||
| AK308159.1 | ||||
| BC009712.2 | AAH09712.1 | |||
| BC068509.1 | AAH68509.1 | |||
| BT006644.1 | AAP35290.1 | |||
| DC317792.1 | ||||
| M81182.1 | AAA60128.1 | |||
| X58528.1 | CAA41416.1 |
More Information
Set Membership: |
|

Genomic Map