Assay Details
Target Gene Details
Entrez Gene ID: | 10236 |
Gene Name: | heterogeneous nuclear ribonucleoprotein R |
Gene Aliases: |
HNRPR, hnRNP-R |
Location: |
Chr.1:23304688-23344650 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
More Information
Set Membership: |
|

Genomic Map