Assay Details
Target Gene Details
Entrez Gene ID: | 57795 |
Gene Name: | BMP/retinoic acid inducible neural specific 2 |
Gene Aliases: |
DBCCR1L2, FAM5B |
Location: |
Chr.1:177170922-177282422 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BRINP2 | XM_005245379.1 | 1 | 315 | XP_005245436.1 |
| AK296524.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs146812772] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map