Assay Details
Target Gene Details
Entrez Gene ID: | 101929592 |
Gene Name: | uncharacterized LOC101929592 |
Gene Aliases: |
- |
Location: |
Chr.1:43709392-43727343 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929592 | NR_125986.1 | |||
| BX280555.1 |
Target Gene Details
Entrez Gene ID: | 6487 |
Gene Name: | ST3 beta-galactoside alpha-2,3-sialyltransferase 3 |
Gene Aliases: |
EIEE15, MRT12, SIAT6, ST3GALII, ST3GalIII, ST3N |
Location: |
Chr.1:43707497-43931165 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
More Information
Additional Information:
For this assay, SNP(s) [rs79570563] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map