Assay Details
Target Gene Details
Entrez Gene ID: | 4985 |
Gene Name: | opioid receptor delta 1 |
Gene Aliases: |
OPRD |
Location: |
Chr.1:28812142-28863696 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OPRD1 | NM_000911.3 | NP_000902.3 | ||
| EF057065.1 | ||||
| EU883570.1 | ||||
| U07882.2 | AAA18789.2 | |||
| U10504.1 | AAA83426.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525310 | Chr.1:28812463 - 28818676 on Build GRCh38 | Loss |
|
| nsv545921 | Chr.1:28781383 - 28839626 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map