Assay Details
Target Gene Details
Entrez Gene ID: | 22977 |
Gene Name: | aldo-keto reductase family 7 member A3 |
Gene Aliases: |
AFAR2 |
Location: |
Chr.1:19274309-19289362 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKR7A3 | NM_012067.2 | NP_036199.2 | ||
| XM_011541046.2 | XP_011539348.1 | |||
| XM_017000714.1 | XP_016856203.1 | |||
| AF040639.1 | AAD02195.1 | |||
| AJ271799.1 | CAC81076.1 | |||
| BC025709.1 | AAH25709.1 | |||
| BC031562.1 | AAH31562.1 | |||
| BC042420.1 | ||||
| BM920315.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3552788 | Chr.1:19266487 - 19285352 on Build GRCh38 | Deletion |
|
| esv26443 | Chr.1:19273138 - 19288033 on Build GRCh38 | Loss |
|
| dgv25e199 | Chr.1:19270091 - 19287665 on Build GRCh38 | Deletion |
|
| nsv945807 | Chr.1:19282774 - 19286791 on Build GRCh38 | Duplication |
|
| esv3585392 | Chr.1:19272870 - 19287742 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map