Assay Details
Target Gene Details
Entrez Gene ID: | 400797 |
Gene Name: | long intergenic non-protein coding RNA 83 |
Gene Aliases: |
NCRNA00083 |
Location: |
Chr.1:178479514-178499623 on Build GRCh38 |
Assay Gene Location: | Overlaps - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00083 | XM_017003086.1 | XP_016858575.1 |
Target Gene Details
Entrez Gene ID: | 9462 |
Gene Name: | RAS protein activator like 2 |
Gene Aliases: |
NGAP |
Location: |
Chr.1:178093729-178479514 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASAL2 | NM_004841.3 | NP_004832.1 | ||
| NM_170692.2 | NP_733793.2 | |||
| AA127237.1 |
More Information
Set Membership: |
|

Genomic Map