Assay Details
Target Gene Details
Entrez Gene ID: | 55532 |
Gene Name: | solute carrier family 30 member 10 |
Gene Aliases: |
HMDPC, HMNDYT1, ZNT10, ZNT8, ZRC1, ZnT-10 |
Location: |
Chr.1:219910398-219959754 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC30A10 | XM_006711437.3 | 4 | 4247 | XP_006711500.2 |
| XM_017001684.1 | 4 | 3498 | XP_016857173.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575695 | Chr.1:219910875 - 219949609 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map