Assay Details
Target Gene Details
Entrez Gene ID: | 10286 |
Gene Name: | breast carcinoma amplified sequence 2 |
Gene Aliases: |
DAM1, SPF27, Snt309 |
Location: |
Chr.1:114567557-114581644 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCAS2 | NM_005872.2 | NP_005863.1 | ||
| AB020623.1 | BAA34863.1 | |||
| AB451245.1 | ||||
| AB451369.1 | ||||
| AF081788.1 | AAC64059.1 | |||
| AK222637.1 | BAD96357.1 | |||
| AK313141.1 | ||||
| BC005285.1 | AAH05285.1 | |||
| BC012623.1 | AAH12623.1 | |||
| BC022880.1 | AAH22880.1 | |||
| BT019390.1 | AAV38197.1 | |||
| CR542037.1 | CAG46834.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv28734 | Chr.1:114376708 - 115005298 on Build GRCh38 | Loss |
|
| nsv831104 | Chr.1:114455501 - 114627235 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map