Assay Details
Target Gene Details
Entrez Gene ID: | 2005 |
Gene Name: | ELK4, ETS transcription factor |
Gene Aliases: |
SAP1 |
Location: |
Chr.1:205597774-205634077 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELK4 | NM_001973.3 | 5 | 7619 | NP_001964.2 |
| AF075044.1 | 1 | 204 | ||
| AK024944.1 | 1 | 544 | ||
| BC040145.1 | AAH40145.1 |
More Information
Set Membership: |
|

Genomic Map