Assay Details
Target Gene Details
Entrez Gene ID: | 27095 |
Gene Name: | trafficking protein particle complex 3 |
Gene Aliases: |
BET3 |
Location: |
Chr.1:36136569-36156053 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRAPPC3 | NM_001270895.1 | NP_001257824.1 | ||
| BM451581.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545995 | Chr.1:36150068 - 36286809 on Build GRCh38 | Loss |
|
| nsv545994 | Chr.1:36147779 - 36244437 on Build GRCh38 | Loss |
|
| nsv545993 | Chr.1:36126011 - 36187018 on Build GRCh38 | Loss |
|
| nsv834691 | Chr.1:36059137 - 36238540 on Build GRCh38 | Loss |
|
| nsv545992 | Chr.1:36091266 - 36161590 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map