Assay Details
Target Gene Details
Entrez Gene ID: | 64757 |
Gene Name: | mitochondrial amidoxime reducing component 1 |
Gene Aliases: |
MOSC1 |
Location: |
Chr.1:220786697-220814399 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MARC1 | NM_022746.3 | NP_073583.3 | ||
| XM_011509900.2 | XP_011508202.1 | |||
| XM_011509903.2 | XP_011508205.1 | |||
| XM_011509904.2 | XP_011508206.1 | |||
| XM_017002096.1 | XP_016857585.1 | |||
| XM_017002097.1 | XP_016857586.1 | |||
| AK000035.1 | ||||
| AK026043.1 | BAB15333.1 | |||
| AK092439.1 | ||||
| AK094105.1 | BAC04286.1 | |||
| AK225416.1 | ||||
| AK290812.1 | ||||
| BC010619.1 | AAH10619.1 |
More Information
Set Membership: |
|

Genomic Map