Assay Details
Target Gene Details
Entrez Gene ID: | 79961 |
Gene Name: | DENN domain containing 2D |
Gene Aliases: |
- |
Location: |
Chr.1:111185969-111204791 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DENND2D | NM_001271833.1 | 12 | 3148 | NP_001258762.1 |
| NM_024901.4 | 12 | 3216 | NP_079177.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv10639 | Chr.1:111126227 - 111244431 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map