Assay Details
Target Gene Details
Entrez Gene ID: | 81573 |
Gene Name: | ankyrin repeat domain 13C |
Gene Aliases: |
dJ677H15.3 |
Location: |
Chr.1:70259002-70354977 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD13C | NM_030816.4 | 13 | 4452 | NP_110443.3 |
| XM_005271234.3 | 12 | 4594 | XP_005271291.1 | |
| XM_017002413.1 | 11 | 3675 | XP_016857902.1 | |
| XM_017002416.1 | 8 | 3421 | XP_016857905.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1014292 | Chr.1:70223532 - 70333382 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map