Assay Details
Target Gene Details
Entrez Gene ID: | 79639 |
Gene Name: | transmembrane protein 53 |
Gene Aliases: |
NET4 |
Location: |
Chr.1:44653247-44674608 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM53 | NM_001300746.1 | NP_001287675.1 | ||
| NM_001300747.1 | NP_001287676.1 | |||
| NM_001300748.1 | NP_001287677.1 | |||
| NM_024587.3 | NP_078863.2 | |||
| XM_011542138.2 | XP_011540440.1 | |||
| AA490195.1 | ||||
| AK026006.1 | BAB15317.1 | |||
| AK225405.1 | ||||
| AK295356.1 | ||||
| AK296548.1 | ||||
| AK310016.1 | ||||
| AL832539.1 | CAI46125.1 | |||
| BC064520.1 | AAH64520.1 | |||
| BM806701.1 | ||||
| CR457348.1 | CAG33629.1 | |||
| HY029536.1 | ||||
| HY108001.1 |

Genomic Map