Assay Details
Target Gene Details
Entrez Gene ID: | 23381 |
Gene Name: | SMG5, nonsense mediated mRNA decay factor |
Gene Aliases: |
EST1B, LPTS-RP1, LPTSRP1, SMG-5 |
Location: |
Chr.1:156249224-156291498 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMG5 | NM_001323614.1 | NP_001310543.1 | ||
| NM_001323615.1 | NP_001310544.1 | |||
| NM_001323616.1 | NP_001310545.1 | |||
| NM_001323617.1 | NP_001310546.1 | |||
| NM_015327.2 | NP_056142.2 | |||
| XM_017000842.1 | XP_016856331.1 | |||
| XM_017000843.1 | XP_016856332.1 | |||
| AB029012.2 | ||||
| AB085691.1 | BAC53623.1 | |||
| AK027486.1 | BAB55147.1 | |||
| AK092183.1 | ||||
| AL137738.1 | CAH56374.1 | |||
| AY168922.1 | AAO17582.1 | |||
| AY336728.1 | AAQ84301.1 | |||
| BC007453.2 | AAH07453.2 | |||
| BC038296.1 | AAH38296.1 | |||
| CR933677.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509524 | Chr.1:156238274 - 156308437 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map