Assay Details
Target Gene Details
Entrez Gene ID: | 54855 |
Gene Name: | family with sequence similarity 46 member C |
Gene Aliases: |
- |
Location: |
Chr.1:117605982-117628389 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM46C | NM_017709.3 | NP_060179.2 | ||
| AK000209.1 | BAA91010.1 | |||
| BC036516.2 | ||||
| BC131726.1 | ||||
| CD300120.1 |
More Information
Set Membership: |
|

Genomic Map