Assay Details
Target Gene Details
Entrez Gene ID: | 348487 |
Gene Name: | family with sequence similarity 131 member C |
Gene Aliases: |
C1orf117 |
Location: |
Chr.1:16057769-16073632 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM131C | NM_182623.2 | NP_872429.2 | ||
| AK094085.1 | BAC04282.1 | |||
| AL529663.3 | ||||
| BC016848.1 | AAH16848.1 | |||
| BC037394.1 | AAH37394.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520148 | Chr.1:16005414 - 16139508 on Build GRCh38 | Loss |
|
| nsv829570 | Chr.1:16032866 - 16077137 on Build GRCh38 | Gain |
|
| nsv508936 | Chr.1:15915852 - 16181025 on Build GRCh38 | Insertion |
|
| nsv833758 | Chr.1:15969028 - 16140519 on Build GRCh38 | Gain+Loss |
|
| nsv954844 | Chr.1:16013606 - 16077905 on Build GRCh38 | Deletion |
|
| nsv470698 | Chr.1:16013965 - 16070311 on Build GRCh38 | Loss |
|
| nsv997887 | Chr.1:15994515 - 16115208 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs141455783] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map