Assay Details
Target Gene Details
Entrez Gene ID: | 9580 |
Gene Name: | SRY-box 13 |
Gene Aliases: |
ICA12, Sox-13 |
Location: |
Chr.1:204073096-204127743 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOX13 | NM_005686.2 | NP_005677.2 | ||
| XM_005245623.2 | XP_005245680.1 | |||
| AF083105.1 | AAC83687.1 | |||
| AF116571.1 | AAF23875.1 | |||
| AK304801.1 | ||||
| AK314197.1 | ||||
| BC106038.1 | AAI06039.1 | |||
| BQ935858.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs115640184] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map