Assay Details
Target Gene Details
Entrez Gene ID: | 9890 |
Gene Name: | phospholipid phosphatase related 4 |
Gene Aliases: |
LPPR4, LPR4, PHP1, PRG-1, PRG1 |
Location: |
Chr.1:99262956-99309584 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLPPR4 | NM_001166252.1 | NP_001159724.1 | ||
| NM_014839.4 | NP_055654.2 | |||
| XM_011542498.2 | XP_011540800.1 | |||
| AB007924.3 | ||||
| AF357013.1 | AAO85400.1 | |||
| AF541281.1 | AAP57770.1 | |||
| AK291369.1 | ||||
| AK300361.1 | ||||
| AL834390.1 | CAD39052.1 | |||
| AY304518.1 | AAP72155.1 | |||
| BC042963.1 | AAH42963.1 |
More Information
Set Membership: |
|

Genomic Map