Assay Details
Target Gene Details
Entrez Gene ID: | 22826 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C8 |
Gene Aliases: |
HSPC331, SPF31 |
Location: |
Chr.1:28200278-28233031 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC8 | NM_014280.2 | NP_055095.2 | ||
| AF083190.1 | AAC35352.1 | |||
| AK300797.1 | ||||
| BC033159.1 | AAH33159.1 | |||
| BX538122.1 | ||||
| CR456900.1 | CAG33181.1 | |||
| DB294321.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545916 | Chr.1:28208722 - 28624350 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map