Assay Details
Target Gene Details
Entrez Gene ID: | 10485 |
Gene Name: | chromosome 1 open reading frame 61 |
Gene Aliases: |
CROC4 |
Location: |
Chr.1:156404252-156456649 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1orf61 | NM_001320453.1 | NP_001307382.1 | ||
| NM_006365.2 | NP_006356.1 | |||
| NR_135260.1 | ||||
| NR_135261.1 | ||||
| NR_135262.1 | ||||
| NR_135263.1 | ||||
| NR_135264.1 | ||||
| XM_011509067.1 | XP_011507369.1 | |||
| XM_011509068.1 | XP_011507370.1 | |||
| XM_017000061.1 | XP_016855550.1 | |||
| XM_017000062.1 | XP_016855551.1 | |||
| XM_017000063.1 | XP_016855552.1 | |||
| XM_017000064.1 | XP_016855553.1 | |||
| XM_017000065.1 | XP_016855554.1 | |||
| XM_017000066.1 | XP_016855555.1 | |||
| AK095626.1 | ||||
| BC050448.1 | AAH50448.1 | |||
| BC059943.1 | AAH59943.1 | |||
| BG716287.1 | ||||
| BI597719.1 | ||||
| BI667681.1 | ||||
| BI668594.1 | ||||
| DN990706.1 | ||||
| HY158402.1 | ||||
| U49857.1 | AAA93029.1 |

Genomic Map