Assay Details
Target Gene Details
Entrez Gene ID: | 2140 |
Gene Name: | EYA transcriptional coactivator and phosphatase 3 |
Gene Aliases: |
- |
Location: |
Chr.1:27970344-28088671 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EYA3 | NM_001282561.1 | 17 | 2707 | NP_001269490.1 |
| NM_001282562.1 | 17 | 2765 | NP_001269491.1 | |
| NM_001990.3 | 18 | 2845 | NP_001981.2 | |
| XM_006710449.3 | 19 | 2881 | XP_006710512.1 | |
| XM_011541004.2 | 18 | 2741 | XP_011539306.1 | |
| BX648945.1 | 18 | 2814 |
More Information
Set Membership: |
|

Genomic Map