Assay Details
Target Gene Details
Entrez Gene ID: | 2271 |
Gene Name: | fumarate hydratase |
Gene Aliases: |
FMRD, HLRCC, LRCC, MCL, MCUL1 |
Location: |
Chr.1:241497557-241519785 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FH | NM_000143.3 | NP_000134.2 | ||
| XM_011544132.2 | XP_011542434.1 | |||
| AK312415.1 | ||||
| BC003108.1 | AAH03108.1 | |||
| BC017444.1 | AAH17444.1 | |||
| BT009839.1 | AAP88841.1 | |||
| DA129148.1 | ||||
| KU178071.1 | ||||
| KU178072.1 | ||||
| KU178073.1 | ||||
| KU178074.1 | ||||
| KU178075.1 | ||||
| KU178076.1 | ||||
| M15502.1 | AAA52483.1 | |||
| U48857.1 | AAD00071.1 | |||
| U59309.1 | AAB66354.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv549460 | Chr.1:241331278 - 241669985 on Build GRCh38 | Gain |
|
| nsv1002214 | Chr.1:241295639 - 241936488 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map