Assay Details
Target Gene Details
Entrez Gene ID: | 2899 |
Gene Name: | glutamate ionotropic receptor kainate type subunit 3 |
Gene Aliases: |
EAA5, GLR7, GLUR7, GluK3, GluR7a |
Location: |
Chr.1:36795527-37034243 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRIK3 | NM_000831.3 | 16 | 7672 | NP_000822.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546001 | Chr.1:36692344 - 36841680 on Build GRCh38 | Loss |
|
| nsv1003721 | Chr.1:36716795 - 36849794 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map