Assay Details
Target Gene Details
Entrez Gene ID: | 8574 |
Gene Name: | aldo-keto reductase family 7 member A2 |
Gene Aliases: |
AFAR, AFAR1, AFB1-AR1, AKR7 |
Location: |
Chr.1:19302708-19312146 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKR7A2 | NM_001320979.1 | 6 | 1600 | NP_001307908.1 |
| NM_003689.3 | 7 | 1705 | NP_003680.2 | |
| BG385885.1 | 1 | 279 | ||
| BU944847.1 | 1 | 776 |
More Information
Set Membership: |
|

Genomic Map