Assay Details
Target Gene Details
Entrez Gene ID: | 54558 |
Gene Name: | spermatogenesis associated 6 |
Gene Aliases: |
HASH, SRF-1, SRF1 |
Location: |
Chr.1:48258643-48472208 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPATA6 | NM_001286238.1 | NP_001273167.1 | ||
| NM_001286239.1 | NP_001273168.1 | |||
| NM_019073.3 | NP_061946.1 | |||
| XM_006710699.2 | XP_006710762.1 | |||
| XM_006710700.2 | XP_006710763.1 | |||
| XM_011541606.1 | XP_011539908.1 | |||
| XM_011541607.2 | XP_011539909.1 | |||
| XM_011541608.1 | XP_011539910.1 | |||
| XM_011541609.2 | XP_011539911.1 | |||
| XM_017001510.1 | XP_016856999.1 | |||
| XM_017001511.1 | XP_016857000.1 | |||
| XM_017001512.1 | XP_016857001.1 | |||
| XM_017001513.1 | XP_016857002.1 | |||
| XM_017001514.1 | XP_016857003.1 | |||
| XM_017001515.1 | XP_016857004.1 | |||
| AK000869.1 | BAA91403.1 | |||
| AK301770.1 | ||||
| BC020660.1 | AAH20660.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3585965 | Chr.1:48282463 - 48313854 on Build GRCh38 | Loss |
|
| esv2762139 | Chr.1:48279401 - 48310271 on Build GRCh38 | Loss |
|
| nsv1011588 | Chr.1:48248029 - 48328196 on Build GRCh38 | Gain |
|
| esv3578249 | Chr.1:48202315 - 48332128 on Build GRCh38 | Gain |
|
| nsv508182 | Chr.1:48264259 - 48353795 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs112008608,rs72891535] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map