Assay Details
Target Gene Details
Entrez Gene ID: | 9829 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C6 |
Gene Aliases: |
DJC6, PARK19 |
Location: |
Chr.1:65264694-65415869 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC6 | NM_001256865.1 | NP_001243794.1 | ||
| NM_014787.3 | NP_055602.1 | |||
| AB007942.1 | ||||
| AK296408.1 | ||||
| BC051722.1 | AAH51722.1 | |||
| BP311649.1 | ||||
| DC332732.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2676423 | Chr.1:64977641 - 65405798 on Build GRCh38 | Deletion |
|
| esv3586278 | Chr.1:65283752 - 65299660 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map