Assay Details
Target Gene Details
Entrez Gene ID: | 6924 |
Gene Name: | transcription elongation factor B subunit 3 |
Gene Aliases: |
EloA, SIII, SIII p110, TCEB3A |
Location: |
Chr.1:23743366-23762059 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCEB3 | NM_003198.2 | 11 | 4231 | NP_003189.2 |
| AF070554.1 | 1 | 950 |
Target Gene Details
Entrez Gene ID: | 100506963 |
Gene Name: | TCEB3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:23760382-23778297 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCEB3-AS1 | NR_038280.1 | |||
| AL049675.1 | ||||
| BG429664.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs77646517] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map