Assay Details
Target Gene Details
Entrez Gene ID: | 3738 |
Gene Name: | potassium voltage-gated channel subfamily A member 3 |
Gene Aliases: |
HGK5, HLK3, HPCN3, HUKIII, KV1.3, MK3, PCN3 |
Location: |
Chr.1:110653560-110675033 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNA3 | NR_109845.1 | |||
| NR_109846.1 | ||||
| DB086905.1 | ||||
| HY035404.1 |
More Information
Set Membership: |
|

Genomic Map